Assay Design



Gene Transcripts

Gene Symbol
RPUSD4
Entrez Gene ID
516934
Gene Name
RNA pseudouridine synthase D4
Gene Aliases
-
Chromosome Location
Chr.29: 29552504 - 29561873 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015616.1 NP_001015616.1 6 - 7 904 96
GenBank mRNA BC134590.1 - 6 - 7 926 96
BT020717.1 - 6 - 7 904 96
BT020773.1 - 6 - 7 905 96
BT020778.1 - 6 - 7 904 96
BT021642.1 - 6 - 7 903 96
BT025340.1 - 6 - 7 890 96
BT025483.1 - 6 - 7 896 96
BT026244.1 - 6 - 7 915 96

View Full Product Details