Assay Design



Gene Transcripts

Gene Symbol
MAGOH
Entrez Gene ID
505417
Gene Name
mago homolog, exon junction complex subunit
Gene Aliases
-
Chromosome Location
Chr.3: 93003066 - 93012323 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034245.2 NP_001029417.1 1 - 2 188 135
GenBank mRNA BC103092.1 - 1 - 2 189 135

View Full Product Details