Assay Design



Gene Transcripts

Gene Symbol
APOM
Entrez Gene ID
505830
Gene Name
apolipoprotein M
Gene Aliases
-
Chromosome Location
Chr.23: 27641577 - 27646567 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034257.2 NP_001029429.2 4 - 5 525 78
GenBank mRNA BC103165.1 - 4 - 5 527 78
DV807269.1 - 4 - 5 513 78

View Full Product Details