Assay Design



Gene Transcripts

Gene Symbol
TSPAN13
Entrez Gene ID
526460
Gene Name
tetraspanin 13
Gene Aliases
-
Chromosome Location
Chr.4: 25245149 - 25283286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035362.1 NP_001030439.1 3 - 4 496 69
GenBank mRNA BC103095.1 - 3 - 4 496 69

View Full Product Details