Assay Design



Gene Transcripts

Gene Symbol
CA4
Entrez Gene ID
280741
Gene Name
carbonic anhydrase 4
Gene Aliases
-
Chromosome Location
Chr.19: 12804061 - 12811833 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173897.1 NP_776322.1 7 - 8 777 102
GenBank mRNA BC142534.1 - 7 - 8 792 102
U58870.1 - 7 - 8 777 102

View Full Product Details