Assay Design
Gene Transcripts
- Gene Symbol
- GAST
- Entrez Gene ID
-
280800
- Gene Name
- gastrin
- Gene Aliases
-
GAS
- Chromosome Location
-
Chr.19: 41926788 - 41929100 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173915.2 |
NP_776340.1 |
1 - 2 |
50 |
89 |
GenBank mRNA |
X16581.1 |
- |
- |
50 |
89 |
View Full Product Details