Assay Design



Gene Transcripts

Gene Symbol
TTR
Entrez Gene ID
280948
Gene Name
transthyretin
Gene Aliases
-
Chromosome Location
Chr.24: 25544099 - 25552959 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173967.3 NP_776392.1 3 - 4 346 56
GenBank mRNA AB009591.1 - 3 - 4 346 56
BC103035.1 - 3 - 4 363 56

View Full Product Details