Assay Design



Gene Transcripts

Gene Symbol
NPPC
Entrez Gene ID
281356
Gene Name
natriuretic peptide C
Gene Aliases
-
Chromosome Location
Chr.2: 119585622 - 119590237 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174125.2 NP_776550.1 1 - 2 227 81
GenBank mRNA BC123399.1 - 1 - 2 135 81
Z48477.1 - - 227 81
Z48478.1 - - 227 81

View Full Product Details