Assay Design
Gene Transcripts
- Gene Symbol
- PIM1
- Entrez Gene ID
-
281402
- Gene Name
- Pim-1 proto-oncogene, serine/threonine kinase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.23: 11066555 - 11071948 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174144.2 |
NP_776569.1 |
3 - 4 |
646 |
92 |
GenBank mRNA |
AF259078.1 |
- |
3 - 4 |
646 |
92 |
View Full Product Details