Assay Design



Gene Transcripts

Gene Symbol
PRM1
Entrez Gene ID
281423
Gene Name
protamine 1
Gene Aliases
-
Chromosome Location
Chr.25: 9936642 - 9937121 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174156.2 NP_776581.1 1 - 2 185 107
GenBank mRNA M14559.1 - 1 - 2 118 107
M18625.1 - 1 - 2 185 107

View Full Product Details