Assay Design
Gene Transcripts
- Gene Symbol
- NPVF
- Entrez Gene ID
-
281451
- Gene Name
- neuropeptide VF precursor
- Gene Aliases
-
RFRP
- Chromosome Location
-
Chr.4: 70667240 - 70670595 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174168.1 |
NP_776593.1 |
2 - 3 |
537 |
80 |
GenBank mRNA |
AB040291.1 |
- |
2 - 3 |
537 |
80 |
View Full Product Details