Assay Design



Gene Transcripts

Gene Symbol
ATP5PO
Entrez Gene ID
281640
Gene Name
ATP synthase peripheral stalk subunit OSCP
Gene Aliases
ATP5O, OSCP
Chromosome Location
Chr.1: 1643680 - 1651038 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174244.1 NP_776669.1 6 - 7 604 85
GenBank mRNA AB098834.1 - 1 - 2 178 85
BC102204.1 - 6 - 7 565 85
M18753.1 - 6 - 7 604 85

View Full Product Details