Assay Design
Gene Transcripts
- Gene Symbol
- CABP2
- Entrez Gene ID
-
281654
- Gene Name
- calcium binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.29: 45396976 - 45401741 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174255.3 |
NP_776680.2 |
2 - 3 |
153 |
80 |
GenBank mRNA |
AF169155.1 |
- |
2 - 3 |
153 |
80 |
View Full Product Details