Assay Design



Gene Transcripts

Gene Symbol
GNAI1
Entrez Gene ID
281790
Gene Name
G protein subunit alpha i1
Gene Aliases
-
Chromosome Location
Chr.4: 40955829 - 41061584 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174324.2 NP_776749.1 3 - 4 423 120
GenBank mRNA BC105419.1 - 4 - 5 465 120
X03642.1 - - 423 120

View Full Product Details