Assay Design



Gene Transcripts

Gene Symbol
HSPE1
Entrez Gene ID
281833
Gene Name
heat shock protein family E (Hsp10) member 1
Gene Aliases
-
Chromosome Location
Chr.2: 86046023 - 86048063 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174346.2 NP_776771.1 2 - 3 208 118
GenBank mRNA BC102684.1 - 2 - 3 216 118
X69556.1 - - 208 118

View Full Product Details