Assay Design



Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
281856
Gene Name
interleukin 12A
Gene Aliases
IL12p35, Il-12 p35
Chromosome Location
Chr.1: 107433482 - 107440180 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174355.2 NP_776780.1 3 - 4 278 96
GenBank mRNA EU276075.1 - 3 - 4 278 96
U14416.1 - 3 - 4 278 96

View Full Product Details