Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174378.2 | NP_776803.1 | 2 - 3 | 320 | 121 |
GenBank mRNA | BC102173.1 | - | 2 - 3 | 320 | 121 |
BT025469.1 | - | 2 - 3 | 304 | 121 | |
FJ232912.1 | - | 2 - 3 | 293 | 121 | |
J05147.1 | - | 2 - 3 | 320 | 121 | |
M18780.1 | - | 2 - 3 | 301 | 121 |