Assay Design



Gene Transcripts

Gene Symbol
FCGR2B
Entrez Gene ID
282229
Gene Name
Fc fragment of IgG receptor IIb
Gene Aliases
CD32, FCGR2, fcRII
Chromosome Location
Chr.3: 7931199 - 7947831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174539.2 NP_776964.1 6 - 7 885 61
GenBank mRNA BC113215.1 - 6 - 7 873 61
DQ868526.1 - 6 - 7 998 61
EU151550.1 - 5 - 6 686 61
EU151552.1 - 4 - 5 416 61
X75671.1 - - 885 61

View Full Product Details