Assay Design
Gene Transcripts
- Gene Symbol
- CYP11B1
- Entrez Gene ID
-
282422
- Gene Name
- cytochrome P450, subfamily XI B, polypeptide 1
- Gene Aliases
-
P450c11
- Chromosome Location
-
Chr.14: 1550747 - 1558462 on Build ARS-UCD1.2
- Gene Symbol
- LOC787647
- Entrez Gene ID
-
787647
- Gene Name
- cytochrome P450 11B1, mitochondrial-like
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D00361.1 |
- |
- |
807 |
66 |
View Full Product Details