Assay Design



Gene Transcripts

Gene Symbol
MSX1
Entrez Gene ID
286872
Gene Name
msh homeobox 1
Gene Aliases
-
Chromosome Location
Chr.6: 104281517 - 104285789 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174798.2 NP_777223.1 1 - 2 690 94
GenBank mRNA BC120208.1 - 1 - 2 703 94
D30750.1 - 1 - 2 690 94

View Full Product Details