Assay Design



Gene Transcripts

Gene Symbol
NDUFA1
Entrez Gene ID
327673
Gene Name
NADH:ubiquinone oxidoreductase subunit A1
Gene Aliases
CI-MWFE, MWFE
Chromosome Location
Chr.X: 4220541 - 4223723 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175794.2 NP_786988.1 1 - 2 159 93
GenBank mRNA BC102993.1 - 1 - 2 168 93
X63222.1 - - 159 93

View Full Product Details