Assay Design



Gene Transcripts

Gene Symbol
NDUFA2
Entrez Gene ID
327698
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases
B8, CI-B8
Chromosome Location
Chr.7: 51778419 - 51780743 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175815.2 NP_787009.1 2 - 3 243 103
GenBank mRNA BC118428.1 - 2 - 3 230 103
MK423999.1 - 2 - 3 153 103
X63219.1 - - 243 103

View Full Product Details