Assay Design
Gene Transcripts
- Gene Symbol
- PAG11
- Entrez Gene ID
-
337907
- Gene Name
- pregnancy-associated glycoprotein 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.29: 39438824 - 39456485 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176623.2 |
NP_788796.1 |
5 - 6 |
639 |
91 |
GenBank mRNA |
AF020513.1 |
- |
5 - 6 |
639 |
91 |
View Full Product Details