Assay Design



Gene Transcripts

Gene Symbol
UMPS
Entrez Gene ID
281568
Gene Name
uridine monophosphate synthetase
Gene Aliases
-
Chromosome Location
Chr.1: 69127829 - 69177876 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177508.1 NP_803474.1 5 - 6 1308 100
GenBank mRNA BC112872.1 - 5 - 6 1306 100
X65125.1 - - 1308 100

View Full Product Details