Assay Design
Gene Transcripts
- Gene Symbol
- AANAT
- Entrez Gene ID
-
281583
- Gene Name
- aralkylamine N-acetyltransferase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 55288452 - 55290293 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177509.2 |
NP_803475.1 |
2 - 3 |
270 |
54 |
GenBank mRNA |
AD000742.1 |
- |
2 - 3 |
270 |
54 |
View Full Product Details