Assay Design



Gene Transcripts

Gene Symbol
FOXH1
Entrez Gene ID
493733
Gene Name
forkhead box H1
Gene Aliases
FAST1
Chromosome Location
Chr.14: 469132 - 470687 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008414.2 NP_001008414.2 2 - 3 363 89

View Full Product Details