Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012682.1 | NP_001012700.1 | 3 - 4 | 285 | 75 |
GenBank mRNA | AB098956.1 | - | 2 - 3 | 285 | 75 |
AB098998.1 | - | 2 - 3 | 295 | 75 | |
AF013214.1 | - | 1 - 2 | 178 | 75 | |
BC102074.1 | - | 3 - 4 | 299 | 75 | |
BC151695.1 | - | 3 - 4 | 297 | 75 | |
BT021080.1 | - | 3 - 4 | 285 | 75 |