Assay Design



Gene Transcripts

Gene Symbol
COX6B2
Entrez Gene ID
503554
Gene Name
cytochrome c oxidase subunit 6B2
Gene Aliases
-
Chromosome Location
Chr.18: 62124752 - 62126265 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012684.1 NP_001012702.1 1 - 2 328 59
GenBank mRNA AY152401.1 - 1 - 2 328 59
BC109911.1 - 1 - 2 304 59

View Full Product Details