Assay Design



Gene Transcripts

Gene Symbol
CEL
Entrez Gene ID
280748
Gene Name
carboxyl ester lipase
Gene Aliases
-
Chromosome Location
Chr.11: 103058913 - 103067774 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013583.1 NP_001013601.1 10 - 11 1495 100
GenBank mRNA BC149530.1 - 10 - 11 1505 100
BC149638.1 - 10 - 11 1502 100
BT021633.1 - 10 - 11 1495 100
M28402.1 - 10 - 11 1477 100

View Full Product Details