Assay Design



Gene Transcripts

Gene Symbol
CEL
Entrez Gene ID
280748
Gene Name
carboxyl ester lipase
Gene Aliases
-
Chromosome Location
Chr.11: 103058913 - 103067774 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013583.1 NP_001013601.1 1 - 2 74 103
GenBank mRNA BC149530.1 - 1 - 2 84 103
BC149638.1 - 1 - 2 81 103
BT021633.1 - 1 - 2 74 103
M28402.1 - 1 - 2 56 105

View Full Product Details