Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014388.1 | NP_001014410.1 | 3 - 4 | 394 | 122 |
GenBank mRNA | AB098973.1 | - | 3 - 4 | 384 | 123 |
AB099031.1 | - | 3 - 4 | 398 | 122 | |
BC104562.1 | - | 3 - 4 | 403 | 122 | |
BT021036.1 | - | 3 - 4 | 394 | 122 |