Assay Design



Gene Transcripts

Gene Symbol
CNPPD1
Entrez Gene ID
507473
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2H2orf24, C2orf24
Chromosome Location
Chr.2: 107121224 - 107126932 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015538.1 NP_001015538.1 5 - 6 585 79
GenBank mRNA BC113239.1 - 5 - 6 601 79
BT020928.1 - 5 - 6 585 79

View Full Product Details