Assay Design



Gene Transcripts

Gene Symbol
SFXN1
Entrez Gene ID
511375
Gene Name
sideroflexin 1
Gene Aliases
-
Chromosome Location
Chr.10: 5614077 - 5675275 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015574.1 NP_001015574.1 1 - 2 68 114
GenBank mRNA BC103181.1 - 1 - 2 82 114
BT020892.1 - 1 - 2 68 114

View Full Product Details