Assay Design



Gene Transcripts

Gene Symbol
RAE1
Entrez Gene ID
513080
Gene Name
ribonucleic acid export 1
Gene Aliases
-
Chromosome Location
Chr.13: 58787475 - 58805839 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015585.1 NP_001015585.1 10 - 11 1016 64
GenBank mRNA BC111248.1 - 10 - 11 1013 64
BT021016.1 - 10 - 11 1016 64

View Full Product Details