Assay Design



Gene Transcripts

Gene Symbol
SLC25A21
Entrez Gene ID
513423
Gene Name
solute carrier family 25 member 21
Gene Aliases
-
Chromosome Location
Chr.21: 46880360 - 47401047 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015587.1 NP_001015587.1 6 - 7 686 79
GenBank mRNA BT020902.1 - 6 - 7 686 79

View Full Product Details