Assay Design



Gene Transcripts

Gene Symbol
EIF2B2
Entrez Gene ID
513958
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
-
Chromosome Location
Chr.10: 86259480 - 86266557 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015593.1 NP_001015593.1 1 - 2 193 68
GenBank mRNA BC123823.1 - 1 - 2 197 68
BT021006.1 - 1 - 2 193 68

View Full Product Details