Assay Design



Gene Transcripts

Gene Symbol
GSTA4
Entrez Gene ID
533917
Gene Name
glutathione S-transferase alpha 4
Gene Aliases
-
Chromosome Location
Chr.23: 25209828 - 25224023 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015651.1 NP_001015651.1 3 - 4 306 68
GenBank mRNA BC109801.1 - 4 - 5 333 68
BT020960.1 - 3 - 4 306 68

View Full Product Details