Assay Design



Gene Transcripts

Gene Symbol
CORO1B
Entrez Gene ID
510128
Gene Name
coronin 1B
Gene Aliases
-
Chromosome Location
Chr.29: 45329804 - 45335272 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113243.1 NP_001106714.1 7 - 8 939 84
GenBank mRNA BC103376.1 - 7 - 8 921 84
BT021806.1 - 7 - 8 909 84

View Full Product Details