Assay Design



Gene Transcripts

Gene Symbol
KCND1
Entrez Gene ID
518384
Gene Name
potassium voltage-gated channel subfamily D member 1
Gene Aliases
-
Chromosome Location
Chr.X: 87068162 - 87076835 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024553.1 NP_001019724.1 2 - 3 1419 70
GenBank mRNA AY989810.1 - 2 - 3 1419 70

View Full Product Details