Assay Design



Gene Transcripts

Gene Symbol
ELF5
Entrez Gene ID
539420
Gene Name
E74 like ETS transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.15: 65019518 - 65052738 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024569.1 NP_001019740.1 3 - 4 483 94
GenBank mRNA BC151526.1 - 3 - 4 469 94
BT021517.1 - 3 - 4 483 94
DQ126155.1 - 2 - 3 314 94

View Full Product Details