Assay Design



Gene Transcripts

Gene Symbol
OAZ1
Entrez Gene ID
506973
Gene Name
ornithine decarboxylase antizyme 1
Gene Aliases
-
Chromosome Location
Chr.7: 21282787 - 21286350 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127243.2 NP_001120715.1 3 - 4 574 118
GenBank mRNA AY911315.1 - 3 - 4 550 118
BC105187.1 - 3 - 4 543 118
BC146117.1 - 3 - 4 555 118
DV821888.1 - 3 - 4 547 118
EV679539.1 - 3 - 4 577 118

View Full Product Details