Assay Design



Gene Transcripts

Gene Symbol
CLCN4
Entrez Gene ID
511699
Gene Name
chloride voltage-gated channel 4
Gene Aliases
-
Chromosome Location
Chr.X: 132545378 - 132617272 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031757.1 NP_001026927.1 3 - 4 429 79
GenBank mRNA BC151324.1 - 2 - 3 208 79
DQ102705.1 - 3 - 4 429 79

View Full Product Details