Assay Design



Gene Transcripts

Gene Symbol
EDC3
Entrez Gene ID
517979
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16
Chromosome Location
Chr.21: 33991582 - 34046897 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031758.1 NP_001026928.1 5 - 6 1115 79
GenBank mRNA BT021539.1 - 5 - 6 1115 79

View Full Product Details