Assay Design



Gene Transcripts

Gene Symbol
MFF
Entrez Gene ID
506291
Gene Name
mitochondrial fission factor
Gene Aliases
C2H2orf33, C2orf33
Chromosome Location
Chr.2: 115486522 - 115515501 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034267.2 NP_001029439.1 5 - 6 659 114
GenBank mRNA BC102501.1 - 5 - 6 660 114

View Full Product Details