Assay Design



Gene Transcripts

Gene Symbol
YIPF5
Entrez Gene ID
521829
Gene Name
Yip1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.7: 55257073 - 55275266 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034543.1 NP_001029715.1 2 - 3 240 80
GenBank mRNA BC102241.1 - 2 - 3 240 80
BT020972.1 - 2 - 3 201 80
BT029906.1 - 2 - 3 178 80

View Full Product Details