Assay Design



Gene Transcripts

Gene Symbol
DEDD
Entrez Gene ID
538428
Gene Name
death effector domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 8363029 - 8374025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034643.2 NP_001029815.1 1 - 2 43 65
GenBank mRNA BC102576.1 - 1 - 2 44 65

View Full Product Details