Assay Design



Gene Transcripts

Gene Symbol
RWDD1
Entrez Gene ID
541216
Gene Name
RWD domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 34136376 - 34154203 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034726.2 NP_001029898.1 6 - 7 712 139
GenBank mRNA BC102910.1 - 6 - 7 713 139

View Full Product Details