Assay Design



Gene Transcripts

Gene Symbol
MIF4GD
Entrez Gene ID
613305
Gene Name
MIF4G domain containing
Gene Aliases
-
Chromosome Location
Chr.19: 56218574 - 56223189 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034732.1 NP_001029904.1 5 - 6 584 118
GenBank mRNA BC102971.1 - 5 - 6 584 118

View Full Product Details