Assay Design



Gene Transcripts

Gene Symbol
DAD1
Entrez Gene ID
614538
Gene Name
defender against cell death 1
Gene Aliases
-
Chromosome Location
Chr.10: 22229392 - 22251343 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034761.1 NP_001029933.1 2 - 3 452 107
GenBank mRNA BC102081.1 - 2 - 3 440 107
BT020998.1 - 2 - 3 452 107

View Full Product Details