Assay Design



Gene Transcripts

Gene Symbol
NXPH1
Entrez Gene ID
614571
Gene Name
neurexophilin 1
Gene Aliases
-
Chromosome Location
Chr.4: 16522990 - 16891354 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034762.1 NP_001029934.1 2 - 3 585 110
GenBank mRNA BC123618.1 - 2 - 3 423 110
BT020894.1 - 2 - 3 585 110

View Full Product Details